Would you like to log in again?
ALS Human ELISA, BioVendor
Supplier: BioVendor
E35
102552-050EA 938.31 USD InStock
102552-050
ALS Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of ALS.
  • Calibration Range: 0 to 40 µM/ml
  • Limit of Detection: 0.23 mU/ml
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 10 µl/well
The Insulin-like Growth Factors (IGF) – I and II are bound to specific binding proteins in circulation (IGFBP). Until today seven different proteins have been identified IGFBP-1 to 7. IGF bioavailablity, transport and storage is regulated or facilitated by these binding proteins which are expressed differentially according physiological and developmental requirements. The most abundant IGFBP in circulation is IGFBP-3. Together with IGFBP-5 it is able to form the so called ternary complex with IGF and the acid-labile subunit (ALS). In the circulation nearly all IGF is bound in this ternary complex and thus not able to cross the endothelial barrier. Only very small amounts of IGF or IGFBP-3 exist outside this complex. The acid-labile subunit is an important part of the IGF-storage mechanism in circulation. In ALS deficiency or in ALS knock-out mice the concentration of IGF and IGFBP-3 in the circulation is significantly decreased resulting in impaired growth. The acid-labile Subunit, is a synthezised as propeptide of 605 amino acids. The signal peptide, necessary for ALS secretion (AA 1–27) cleaved off enduring the transport process (Swiss-Prot P35858 Version 82). The mature protein consists of 578 amino acids and contains about 20 leucin rich sequence repeats. Beside the leucin-rich repeats several potential N-linked glycosylation sides have been described. Miller BS et al. were able to demonstrate that incomplete glycolsylation of IGFs, ALS and IGFBP-3 results in a decreased serum concentration of these proteins. Oral mannose therapy resulted in a partial normalization of the glycosylation pattern and went along with improved growth. Mutations in or the complete knock out of the ALS gene result in IGF / IGFBP-3 deficiency and therewith in disturbation of growth. Beside growth also other endocrine axes may be involved. In primary ALS deficiency hyperinsulinemia could be observed. Further, the HGH-IGF-IGFBP-system seems to be of relevance in coronary disease]. The first ALS immunoassay was described by Baxter RC in 1990 [6]. By this in-house radioimmunoassay it was shown that ALS is present in high concentrations in serum (50μg/mL) of healthy humans. But not detectable in other body fluids like amniotic fluid, cerebrospinal fluid or seminal plasma – in spite of the fact that these body fluids contain high level IGFBP-3.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty