Would you like to log in again?
Albumin Urinary Rat ELISA, BioVendor
Supplier: BioVendor
638-25309
102552-060EA 843.15 USD InStock
102552-060
Albumin Urinary Rat ELISA, BioVendor
Assays ELISAs
Albumin refers generally to any protein with water solubility
  • Calibration Range: 6.17–500 ug/mL
  • Plasma, Serum, Urine: 7 uL/well
Albumin refers generally to any protein with water solubility, which is moderately soluble in concentrated salt solutions, and experiences heat coagulation (protein denaturation). The most well-known type of albumin is the serum albumin in the blood. Serum albumin is the most abundant blood plasma protein and is produced in the liver.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty