Would you like to log in again?
Alpha-1-Antitrypsin in Stool, A1AT, Human ELISA, BioVendor
Supplier: BioVendor
IC6200
102552-066EA 926.26 USD InStock
102552-066
Alpha-1-Antitrypsin in Stool, A1AT, Human ELISA, BioVendor
Assays ELISAs
The measurement of α-1-AT in stool reflects the permeability of the gut during inflammatory processes.
  • Calibration Range: 3.3-90 ng/mL
  • Limit of Detection: 1.5 ng/mL
  • Plasma-EDTA, Serum, Stool: 100 uL/well
Alpha-1-antitrypsin (α-1-AT) is a 52 kDa glycoprotein, which is produced by the liver, intestinal macrophages, monocytes and mucous membrane cells of the gut. It belongs to the group of acute phase proteins and is one of the most important proteinase inhibitor. α-1-AT inhibits, beside others, the proteinases trypsin and the elastase of neutrophiles. A lack of α-1-AT leads to an enhanced proteolysis. Only a very small amount of α-1-AT is cleaved or resorbed in the gut. Therefore the measurement of α-1-AT in stool reflects the permeability of the gut during inflammatory processes.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty