Would you like to log in again?
Alpha-Fetoprotein Human ELISA, BioVendor
Supplier: BioVendor
RIS0018R
102552-068EA 764.25 USD InStock
102552-068
Alpha-Fetoprotein Human ELISA, BioVendor
Assays ELISAs
AFP is normally produced during fetal and neonatal development by the liver, yolk sac, and in small concentrations by the gastrointestinal tract
  • Calibration Range: 10–160 IU/mL
  • Limit of Detection: 1 IU/mL
  • Serum: 25 uL/well
Alpha-fetoprotein (AFP) is a glycoprotein with a molecular weight of approximately 70 KD. AFP is normally produced during fetal and neonatal development by the liver, yolk sac, and in small concentrations by the gastrointestinal tract. After birth, serum AFP concentrations decrease rapidly, and by the second year of life and thereafter only trace amounts are normally detected in serum. Elevation of serum AFP to abnormally high values occurs in several malignant diseases, most notably nonseminomatous testicular cancer and primary hepatocelluar carcinoma. In the case of nonseminomatous testicular cancer, a direct relationship has been observed between the incidence of elevated AFP levels and the stage of disease. Elevated AFP levels have also been observed in patients diagnosed with seminoma with nonseminomatous elements, but not in patients with pure seminoma. In addition, elevated serum AFP concentrations have been measured in patients with other noncancerous diseases, including ataxia telangiectasia, hereditary tyrosinemia, neonatal hyperbilirubinemia, acute viral hepatitis, chronic active hepatitis and cirrhosis. Elevated serum AFP concentrations are also observed in pregnant women. Therefore, AFP measurements are not recommended for use as a screening procedure to detect the presence of cancer in the general population.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty