Would you like to log in again?
Human Anti IgA isotype IgG ELISA, Biovendor
Supplier: BioVendor
RD199145100R
102552-088EA 984.46 USD InStock
102552-088
Human Anti IgA isotype IgG ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of Anti-IgA isotype IgG.
Human polyclonal immunoglobulins A (IgA) are bound to the microplate wells. Calibrators, control sera and samples of sera are pipetted into the wells and incubated. Anti-IgA antibodies present in the samples are bound to immobilized IgA molecules. The unbound substances are removed by the washing step. The specific polyclonal antibody against human immunoglobulin G (IgG) is then added into the wells. This antibody is conjugated with horseradish peroxidase (HRP) and it is linked to the immobilized complex IgA – IgG anti-IgA during incubation.
The unbound conjugate is removed during the subsequent washing step, and then the substrate solution tetramethylbenzidine (TMB) is added into the wells. The enzyme reaction causes the solution in the wells to turn color (blue) and the intensity of the color is directly proportional to the amount of linked IgG anti-IgA antibodies. Development of the color is stopped by an acid stop reagent, and the absorbance of the resulting yellow product is measured. The concentration of IgG antiIgA antibodies in unknown samples is determined from the calibration curve, which is constructed by plotting absorbance values of calibrators against their known concentration.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty