Would you like to log in again?
sAPO-1/Fas Human ELISA, BioVendor
Supplier: BioVendor
RAF085R
102552-090EA 1054.98 USD InStock
102552-090
sAPO-1/Fas Human ELISA, BioVendor
Assays ELISAs
APO-1 also occurs in a soluble form (sAPO-1) devoid of a transmembrane region.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 13.2 pg/mL
  • Cell culture supernatant, Serum: 10 uL/well
Programmed cell death or apoptosis is the most common form of eukaryotic cell death and is found during tumor regression and embryonic development. In the process of selection and eliminiation of autoreactive B and T cells apoptosis is an important process and therefore a prerequisite for the homeostasis of the immune system. Apoptosis is characterized by changes in cellular morphology (e.g. nuclear condensation, membrane bleeding) and biochemically by rapid induction on DNA fragmentation. APO-1/Fas (CD95), a member of the TNF/NGF receptor superfamily, is a glycosylated 48 kDa surface protein containing a single transmembrane region. APO-1 is expressed on a variety of human B and T cell lines, on many different tumor cells and on various normal human tissues. Triggering of APO-1 by its ligand or by certain anti-APO-1 monoclonal antibodies results in rapid induction of programmed cell death, apoptosis, in susceptible cells. The tissue distribution of APO-1 and of the APO-1 ligand suggests that the APO-1 receptor/ligand system plays an important role in various aspects of mammalian development and especially in the homeostasis of the immune system. Expression of the APO-1 cell surface protein is enhanced by IFN-g and TNF and by activation in lymphocytes. APO-1 also occurs in a soluble form (sAPO-1) devoid of a transmembrane region. Elevated sAPO-1 levels have been reported in sera from patients with high-and low-grade malignant B- and T-cell leukemias and systemic lupus erythematosus. sAPO-1 may prevent cells from undergoing APO-1 ligand induced apoptosis. Hence, secretion of sAPO-1 may provide a mechanism for tumor cells to excape immunosurveillance and may be involved in leukemogenesis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty