Would you like to log in again?
Bcl-2 Human ELISA, BioVendor
Supplier: BioVendor
RAF005R
102552-098EA 1054.98 USD InStock
102552-098
Bcl-2 Human ELISA, BioVendor
Assays ELISAs
Bcl-2 initiates a new gene family involved in the regulation of cell death and survival without affecting cell proliferation.
  • Calibration Range: 0.5-32 ng/mL
  • Limit of Detection: 0.5 ng/mL
  • Cell culture supernatant, Serum, Plasma: 20uL/well
Bcl-2 initiates a new gene family involved in the regulation of cell death and survival without affecting cell proliferation. This precise regulation and maintenance of balance between cell proliferation and cell death in multicellular organisms is critical for tissue homeostasis. Bcl-2, encoded by a proto-oncogene, is an intracellular membrane-associated protein that functions to block programmed cell death. Expression of Bcl-2 has been reported in a wide range of hematopoietic cells, nonneoplastic epithelia and epithelial malignancies. Bcl-2 levels are low or undetectable in endothelial cells. The Bcl-2 gene has a unique function among mammalian oncogenes as a negative regulator of apoptosis. It was first discovered because of its involvement in chromosomal translocations commonly found in lymphomas. Bcl-2 is furthermore associated with stem cells committed to differentiation and morphogenesis. A decrease in Bcl-2 levels leads to cell death by apoptosis. Overexpression of Bcl-2 on the other hand protects cells from death, but it is neither able to immortalize normal cells, nor to cause tumorgenic transformation of immortalized cells. Heterogeneous expression of Bcl-2 in malignancies suggests that the gene is differentially regulated. Furthermore, its expression in association with precancerous lesions suggests a role in the early stage of tumorigenesis. Several homologs of Bcl-2 have recently been discovered, some of which function as inhibitors of cell death and others as promoters of apoptosis that oppose the function of the Bcl-2 protein. Bcl-2 protein plays a critical role in oncogenesis as well as resistance to therapy of cancer. Furthermore prognostic significance of Bcl-2 expression has been shown for several malignancies such as Non-Hodgkin’s Lymphoma, squamous cell carcinomas, breast carcinoma, carcinoma of the gallbladder, and thymoma. Dysregulated Bcl-2 expression has been shown in Multiple Myeloma patients and in acute Myeloid leukemia. Bcl-2 has been suggested as a useful marker for adequate IL-2 therapy e.g. in AIDS patients.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty