Would you like to log in again?
Beta2-Microglobulin Human ELISA, BioVendor
Supplier: BioVendor
RCD003R
102552-100EA 610.74 USD InStock
102552-100
Beta2-Microglobulin Human ELISA, BioVendor
Assays ELISAs
β2-microglobulin (β2-M) is a single polypeptide chain containing 100 amino acids and is found on the surface of nucleated cells.
  • Calibration Range: 0.2-10 mg/L
  • Limit of Detection: 0.1 mg/L
  • Serum: 20 uL/well
β2-microglobulin (β2-M) is a single polypeptide chain containing 100 amino acids and is found on the surface of nucleated cells. β2-M is continuously secreted into the circulatory system and therefore maintains a balanced serum level.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty