Would you like to log in again?
sCD137/4-1BB Human ELISA, BioVendor
Supplier: BioVendor
RAF087R
102552-114EA 1054.98 USD InStock
102552-114
sCD137/4-1BB Human ELISA, BioVendor
Assays ELISAs
The soluble isoforms of CD137, sCD137, are generated by differential splicing and can be detected at enhanced concentrations in sera of patients suffering from various diseases.
  • Calibration Range: 0.8-50 ng/mL
  • Limit of Detection: 0.2 ng/mL
  • Cell culture supernatant, Serum: 50 uL/well
CD137 (ILA/4–1BB) is a member of the tumor necrosis factor receptor family, expressed on activated T lymphocytes. The expression of CD137 is strictly activation dependent in primary cells. Co-stimulation through CD137 enhances T lymphocyte activation and may enable rejection of tumors in vivo. The gene for human CD137 resides on chromosome 1q36, in a cluster of related genes, and this chromosomal region is associated with mutations in several malignancies. Bidirectional signalling has been demonstrated for the CD137 receptor/ ligand system. While cross-linking of CD137 activates T lymphocytes, cross-linking of the CD137 ligand has the opposite effect. This reverse signalling through the CD137 ligand inhibits proliferation of T lymphocytes and induces programmed cell death. Soluble forms of members of the TNF receptor family have been demonstrated for the various members. The soluble isoforms of CD137, sCD137, are generated by differential splicing and can be detected at enhanced concentrations in sera of patients suffering from various diseases. The expression of CD137 isoforms in lymphocytes has been shown and association of sCD137 with activation-induced cell death demonstrated. The levels of sCD137 released by activated lymphocytes have been reported to reflect a negative feed-back control of inflammation in some autoimmune diseases, such as rheumatoid arthritis and multiple sclerosis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty