Would you like to log in again?
sCD152/CTLA-4 Human ELISA, BioVendor
Supplier: BioVendor
RAF088R
102552-118EA 1054.98 USD InStock
102552-118
sCD152/CTLA-4 Human ELISA, BioVendor
Assays ELISAs
A native soluble form of CTLA-4 has been described to be present in human serum.
  • Calibration Range: 0.16-10 ng/mL
  • Limit of Detection: 0.13 ng/mL
  • Amniotic fluid, Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum: 10 uL/well
Cytotoxic T lymphocyte associated gene –4 (CTLA-4) was initially described as a classical type I glycoprotein on the surface of activated T-cells (1). CTLA-4 is a member of the Ig gene superfamily and along with its homologue, CD28, is a B7 binding protein. There is evidence that CTLA-4 is a negative regulator of T-cell activation, ligation of CTLA-4 on the T-cell surface initiates a series of biochemical events that attenuate an ongoing immune response . CTLA-4 knock out mice demonstrate profound polyclonal lymphoproliferative disoders that infiltrate most major organ systems and the animals die a few weeks after birth. The animals have increased levels of IgG which illustrates the role of CTLA-4 on humoral immune responses as well. A role for CTLA-4 in autoimmune disease is suggested by the observations that blockade of B7/CTLA-4 interaction exacerbates animal models of autoimmune disease such as experimental autoimmune encephalomyelitis and diabetes. A search for genetic markers that segregate with Graves‘ disease revealed an association with CTLA-4 polymorphisms. CTLA-4 is further more closely linked to autoimmune thyroid disease (ATD). A native soluble form of CTLA-4 has been described to be present in human serum . A correlation of circulating CTLA-4 and ATD has been shown.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty