Would you like to log in again?
sCD23 Human ELISA, BioVendor
Supplier: BioVendor
RAF089R
102552-122EA 959.2 USD InStock
102552-122
sCD23 Human ELISA, BioVendor
Assays ELISAs
Soluble CD23 is also referred to as Blast-2 and as the low affinity IgE receptor (FCeRII).
  • Calibration Range: 12.5-400 U/mL
  • Limit of Detection: 6.8 U/mL
  • Cell culture supernatant, Plasma, Serum: 100 uL/well
CD23 is described as a 45 kD protein found on the surface of IgM bearing B cells, eosinophils, macrophages and some T and NK cells. It is also found on EBV-transformed B cells. Additionally, a released form has been described. When first released the CD23 molecule is 35 kD; however, this form is quickly cleaved to obtain the more stable, soluble form which is 25 kD in size. Recently the structure of the CD23 molecule was characterized by cloning and sequencing techniques. Soluble CD23 has been shown to be the B cell growth factor (BCGF). Soluble CD23 is also referred to as Blast-2 and as the low affinity IgE receptor (FCeRII). It has been speculated that CD23 may upregulate IgE synthesis in conjunction with T cell promoted interleukin-4; however, the specific physiologic role of this molecule is not yet well understood. Elevated levels of CD23 have been found in research studies of samples from people with B cell-derived Chronic Lymphocytic Leukemia (B-CLL), with Hyper IgE Syndrome and post-Bone Marrow Transplantation samples. CD23 levels may be proven to relate to disease course in Hairy Cell Leukemia (HCL).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty