Would you like to log in again?
sCD28 Human ELISA, BioVendor
Supplier: BioVendor
RAF090R
102552-126EA 1055.11 USD InStock
102552-126
sCD28 Human ELISA, BioVendor
Assays ELISAs
CD28 is structurally very closely related to another molecule expressed in activated T-cells, CTLA-4.
  • Calibration Range: 0.8-50 ng/mL
  • Limit of Detection: 0.18 ng/mL
  • Cell culture supernatant, Serum: 50 uL/well
The cluster of differentiation (CD) antigen CD28 is a 44 kDa, disulphide-bonded, homodimeric glycoprotein, which is constitutively expressed on the surface of the majority of T-cells. CD28 is structurally very closely related to another molecule expressed in activated T-cells, CTLA-4. In particular, the hinge region is completely conserved for both molecules. Also CTLA-4 and CD28 were shown to be very similar at the message and at the gene structure level. The corresponding genes co-map on Human chromosome 2q33. Ligation of CD28 by its counter receptor, B7, expressed on the surface of antigen presenting cells, has been shown to induce signals that, in synergy with those derived from engagement of the T-cell receptor by an antigen bound to a major histocompatibilty complex, enhance proliferation and cytokine production. Manipulation of this interaction can have dramatic effects on the outcome of T-cell activation. Considerable research has been done on the CD28/B7 costimulatory pathway. Blocking thereof results in immunosuppression with implications for the treatment of autoimmune diseases, allergy, organ transplantation and graft versus host disease. Activating the CD28/B7 pathway could be useful for including the immune system to recognize and eliminate tumors that evade the immune system.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty