Would you like to log in again?
sCD44std Human ELISA, BioVendor
Supplier: BioVendor
RAF095R
102552-138EA 1055.11 USD InStock
102552-138
sCD44std Human ELISA, BioVendor
Assays ELISAs
CD44 (85–250 kDa) is a polymorphic glycoprotein.
  • Calibration Range: 0.12-4 ng/mL
  • Limit of Detection: 0.015 ng/mL
  • Amniotic fluid, Cell culture supernatant, Plasma, Serum, Urine: 20 uL/well (1:60 prediluted)
CD44 (85–250 kDa) is a polymorphic glycoprotein. Determination of sCD44std will provide more detailed insight into different pathological modifications during cancer and other diseases.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty