Would you like to log in again?
sCD44var, v6, Human ELISA, BioVendor
Supplier: BioVendor
RAF097R
102552-142EA 1054.98 USD InStock
102552-142
sCD44var, v6, Human ELISA, BioVendor
Assays ELISAs
This CD44-family of transmembrane receptor molecules is derived from a single gene located on chromosome 11.
  • Calibration Range: 0.32-10 ng/mL
  • Limit of Detection: 0.05 ng/mL
  • Amniotic fluid, Body fluids, Cell culture supernatant, Serum, Urine: 20 uL/well (1:20 prediluted)
This CD44-family of transmembrane receptor molecules is derived from a single gene located on chromosome 11. Alternative splicing of the mRNA gives rise to the different isoforms, containing inserts of varying sizes in the extracellular domain of the molecule (exons v2-v10). All CD44 isoforms are variably glycosylated. In contrast to standard CD44 (CD44std) which is almost ubiquitously expressed (18), the variety of CD44 isoforms (CD44var) have a much more restricted distribution, e.g., on keratinocytes (exons v3-v10), ephitelial cells (exons v8-v10), activated lymphocytes and macrophages (exon v6). A splice variant of CD44 (exons v4-v7) confers metastatic behaviour in a rat carcinoma model; aberrant expression of splice variants has been detected on a variety of human tumor cell lines as well as primary and metastatic human tumors, including lymphomas, carcinomas (colon, thyroid, mamma, bladder), and gliomas. Detection of abnormal regulation of CD44 splicing thus could be helpful in cancer diagnosis and disease evaluation. The sCD44var(v6) ELISA detects all circulating CD44 isoforms comprising the sCD44var(v6) sequences.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty