Would you like to log in again?
Human Chemerin ELISA, Biovendor
Supplier: BioVendor
RD191136200R
102552-146EA 735.23 USD InStock
102552-146
Human Chemerin ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of chemerin.
In the Biovendor Human Chemerin ELISA, standards, quality controls and samples are incubated in microtitration wells pre-coated with polyclonal anti-human chemerin antibody. After a 60 min incubation followed by washing, biotin labelled polyclonal anti-human chemerin antibody is added and incubated with the captured chemerin for 60 min. After another washing, streptavidin-HRP conjugate is added. After 30 min incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of chemerin. A standard curve is constructed by plotting absorbance values against chemerin concentrations of standards, and concentrations of unknown samples are determined using this standard curve.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty