Would you like to log in again?
Clusterin Human ELISA, BioVendor
Supplier: BioVendor
RD194034200R
102552-150EA 785.08 USD InStock
102552-150
Clusterin Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of clusterin.
  • Calibration Range: 5 - 160 ng/ml
  • Limit of Detection: 0.5 ng/ml
  • Cerebrospinal fluid, plasma-citrate, plasma-EDTA, plasma-heparin, serum, urine: 5 µl/well
The RD194034200R Human Clusterin ELISA is a sandwich enzyme immunoassay for the quantitative measurement of human clusterin. Clusterin (Apolippoprotein J; SP-40,40; TRPM-2; SGP-2; pADHC-9; CLJ; T64; GP III; XIP8) is a highly conserved disulfide-linked secreted heterodimeric glycoprotein of 75-80 kDa but truncated forms targeted to nucleus have also been identified. The protein is constitutively secreted by a number of cell types including epithelial and neuronal cells and is a major protein in physiological fluids including plasma, milk, urine, cerebrospinal fluid and semen. Due to its wide tissue distribution many diverse physiological functions have been attributed to clusterin including sperm maturation, membrane recycling, lipid transportation, tissue remodelling, complement inhibition and cell-cell or cell-substratum interactions.
In the BioVendor Human Clusterin ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with monoclonal anti-human clusterin antibody. After 60 minutes incubation and washing, biotin labelled second monoclonal anti-human clusterin antibody is added and incubated with captured clusterin for 60 minutes. After another washing, streptavidin-HRP conjugate is added. After 30 minutes incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of clusterin. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty