Would you like to log in again?
Cortisol, Saliva, Human ELISA, BioVendor
Supplier: BioVendor
RCD005R
102552-162EA 598.42 USD InStock
102552-162
Cortisol, Saliva, Human ELISA, BioVendor
Assays ELISAs
Cortisol is the most abundant circulating steroid and the major glucocorticoid secreted by the adrenal cortex.
  • Calibration Range: 1-100 ng/mL
  • Limit of Detection: 1 ng/mL
  • Saliva: 50 uL/well
Cortisol is the most abundant circulating steroid and the major glucocorticoid secreted by the adrenal cortex. Cortisol is physiologically effective in blood pressure maintenance and anti-inflammatory activity. It is also involved in calcium absorption, gluconeogenesis as well as the secretion of gastric acid and pepsin. Measurement of blood cortisol levels can be used as an indicator of adrenal function and the differential diagnosis of Addison's and Cushing's diseases as well as adrenal hyperplasia and carcinoma.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty