Would you like to log in again?
C-Peptide Human ELISA, BioVendor
Supplier: BioVendor
RIS005R
102552-164EA 659.15 USD InStock
102552-164
C-Peptide Human ELISA, BioVendor
Assays ELISAs
The determination of plasma C-Peptide allows an assessment of the endogenous insulin production
  • Calibration Range: 0.05–4.9 pmol/mL
  • Limit of Detection: 0.1 pmol/mL
  • Serum: 100 uL/well
Insulin is synthesized in the beta-cells of the islets of Langerhans as a precursor molecule, proinsulin. In the secretory granules of the beta-cells, proinsulin is cleaved into insulin and into a 31-amino-acid peptide, called the Connecting Peptide or C-Peptide. Insulin and C-Peptide are secreted in equimolar amounts. However, because of its longer half-life, the plasma concentration of C-peptide is higher than that of insulin. The determination of plasma C-Peptide allows an assessment of the endogenous insulin production, even in the presence of exogenous insulin administration or in the presence of circulating anti-insulin antibodies. Moreover, the determination of C-Peptide in urine provides a reliable index of the insulin production when blood sampling is difficult or when an integrated estimation of C-Peptide secretion over a period of several hours is requested.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty