Would you like to log in again?
CRP Human ELISA, BioVendor
Supplier: BioVendor
RAP001
102552-166EA 837.46 USD InStock
102552-166
CRP Human ELISA, BioVendor
Assays ELISAs
CRP is an acute-phase protein, produced exclusively in the liver.
  • Calibration Range: 0–100 ug/mL
  • Limit of Detection: 10 pg/mL
  • Plasma, Serum: 10uL/well
CRP is an acute-phase protein, produced exclusively in the liver. Serum concentration of CRP increases significantly in cases of both infectious and noninfectious inflammation, of tissue damage and necrosis and in the presence of malignant tumours. CRP is present in the active stages of inflammatory disorders like rheumatoid arthritis, ankylosing spondylitis, Reiter’s syndrome, psoriatic arthropathy, systemic lupus erythematosus, polyarteritis, ulcerative colitis and Crohn’s disease. The CRP level increases dramatically following microbial infections, and this can be particularly helpful for the diagnosis and monitoring of bacterial septicemia in neonates and other immunocompromised patients at risk.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty