Would you like to log in again?
CTRP1 Human ELISA, BioVendor
Supplier: BioVendor
RD191153100R
102552-176EA 928.89 USD InStock
102552-176
CTRP1 Human ELISA, BioVendor
Assays ELISAs
CTRP1 (Complement C1q tumor necrosis factor-related protein 1) is a member of the CTRP superfamily and is localized to human chromosome 17.
  • Calibration Range: 3.13-100 ng/mL
  • Limit of Detection: 0.016 ng/mL
  • Plasma-Citrate, Serum: 8 uL/well
CTRP1 (Complement C1q tumor necrosis factor-related protein 1) is a member of the CTRP superfamily and is localized to human chromosome 17. It is a 32 kDa secreted glycoprotein comprised of 281 amino acids encoded by the human gene C1QTN1. This protein is a highly conserved paralogue of adiponectin, containing a cluster of collagenlike repeats, a C-terminal globular ‘C1q-like’ domain and an N-terminal signal peptide sequence followed by a variable region and hence is predicted to be secreted protein. CTRP1 is primarily expressed from cells in the stroma vascular fraction of adipose tissue and is also specifically expressed in the zona glomerulosa of the adrenal cortex and in vascular wall tissue. This protein is ubiquitous in most regions of the human brain and is particularly abundant in the spinal cord, but the pattern of expression and functions of CTRP1 in brain are completely unknown. Expression of CTRP1 (like adiponectin) is induced by PPARg (peroxisome-proliferator-activated receptor γ). The LPS -induced increase in CTRP1 gene expression is found to be mediated by TNF-alpha and IL-1beta. Sequence homology inferred that the CTRP1 cytosolic domain is folded as a collagen-like helix followed by a globular domain. The interaction of the globular domain with the V2R was confirmed by pull-down experiments indicating that this structural motif can also interact with cytosolic proteins. Obesity and the metabolic syndrome are frequently associated with elevated levels of aldosterone and the level of CTRP1 is increased in the blood of hypertensive patients, as aldosterone production was stimulated by CTRP1. It is well known that obesity is the leading cause of hypertension. CTRP1 is stimulating aldosterone production through induction of CYP11B2 gene expression. Therefore, CTRP1 could be a molecular link between obesity and hypertension. Areas of investigation: Energy metabolism and body weight regulation, Metabolic syndrome.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty