Would you like to log in again?
DHEAs Human ELISA, BioVendor
Supplier: BioVendor
RCD008R
102552-194EA 561.43 USD InStock
102552-194
DHEAs Human ELISA, BioVendor
Assays ELISAs
DHEAS is a precursor of testosterone and estrone.
  • Calibration Range: 0.005–10 ug/mL
  • Limit of Detection: 0.005 ug/mL
  • Serum: 50 uL/well
Dehydroepiandrosterone sulfate (DHEAS) is produced by the adrenals and gonads. As a result, the determination of the level of DHEA-S in serum is important in the evaluation of the functional state of these glands. DHEAS is a precursor of testosterone and estrone. Besides the adrenals in females, the ovaries have been shown to be an important source of DHEAS. It has been reported that there is a fluctuation day by day of DHEAS in women during the ovulatory cycle. An abnormal testosterone level in women should be accompanied by the estimation of serum DHEAS. The use of serum testosterone determination in conjunction with Elisa of DHEAS can be used to determine if the source of excess androgen production is ovarian or adrenal.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty