Would you like to log in again?
Dihydrotestosterone, DHT, Human ELISA, BioVendor
Supplier: BioVendor
RCD009R
102552-198EA 593.12 USD InStock
102552-198
Dihydrotestosterone, DHT, Human ELISA, BioVendor
Assays ELISAs
5-dihydrotestosterone (DHT) is a steroid similar to testosterone and androstenedione, which belong to a vlase called androgens.
  • Calibration Range: 25-2500 pg/mL
  • Limit of Detection: 6.0 pg/mL
  • Serum: 50 uL/well
5-dihydrotestosterone (DHT) is a steroid similar to testosterone and androstenedione, which belong to a vlase called androgens. DHT is a C19 steroid and possesses androgenic activity. The bulk of androgen production takes place mainly in the Leydig cells of the testes. Androgens circulate in the blood bound to proteins, especially sex hormone binding globulin (SHBG) and albumin. A trace amount of these steroids circulate in the unbound form in the blood and are referred to as the free fractions. DHT has at least three times the binding affinity for SHBG than testosterone. In males about 70% of DHT is derived from peripheral conversion of testosterone, while in females most of the DHT is derived from androstenedione. The major organ to neutralize androgens is the liver. Therefore in the liver the steroid hormones undergo structural modifications that are generally regarded as prerequisites for their biological inactivation. Some metabolites are formed and some are returned to the circulation.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty