Would you like to log in again?
Estradiol Human ELISA, BioVendor
Supplier: BioVendor
RCD011R
102552-210EA 568.49 USD InStock
102552-210
Estradiol Human ELISA, BioVendor
Assays ELISAs
Estradiol is one of the main components of naturally occurring estrogens and is the major estrogen secreted during the menstrual cycle.
  • Calibration Range: 20-3200 pg/mL
  • Limit of Detection: 10 pg/mL
  • Serum: 50 uL/well
Estradiol is one of the main components of naturally occurring estrogens and is the major estrogen secreted during the menstrual cycle. The serum levels of estradiol are low during the follicular phase rising gradually until about one day before ovulation when a marked rise in the estradiol level occurs (Ovulatory Peak). The estradiol level falls rapidly at, or right after ovulation and is again within the levels of the follicular phase. There is a second rise of estradiol around day 21 of the cycle (Luteal Peak). The levels then decline gradually to the lowest level at the onset of the next menstrual cycle.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty