Would you like to log in again?
Estrone Human ELISA, BioVendor
Supplier: BioVendor
RCD010R
102552-214EA 589.6 USD InStock
102552-214
Estrone Human ELISA, BioVendor
Assays ELISAs
Estrone is a steroid like estriol and estradiol, belonging to the class of estrogens.
  • Calibration Range: 15-2000 pg/mL
  • Limit of Detection: 10 pg/mL
  • Serum: 50 uL/well
Estrone is a steroid like estriol and estradiol, belonging to the class of estrogens. The estrogens are involved in the development of female sex organs and secondary sex characteristics. Before the ovum is fertilized the main action of the estrogens is on the growth and function of the reproductive tract in order to prepare it for the fertilized ovum. During the follicular phase of the menstrual cycle the estrone level shows a slight increase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty