Would you like to log in again?
sFas Ligand Human ELISA, BioVendor
Supplier: BioVendor
RAF100R
102552-218EA 1054.98 USD InStock
102552-218
sFas Ligand Human ELISA, BioVendor
Assays ELISAs
Fas is a type I membrane protein that belongs to the TNF/nerve growth factor receptor family.
  • Calibration Range: 0.16-10 ng/mL
  • Limit of Detection: 0.07 ng/mL
  • Amniotic fluid, Cell culture supernatant, Plasma, Serum: 50 uL/well
Fas is a type I membrane protein that belongs to the TNF/nerve growth factor receptor family. Fas mediates apoptosis, the programmed cell death, when it is cross-linked with specific binding partners. FasL is predominantly expressed on activated T-cells and NK cells; thus FasL-mediated cell death is involved in the T or NK cell-mediated cytotoxicity, some pathologic tissue damages, and the regulation of lymphocyte homeostasis. Elevated expression levels of FasL have been measured in various proliferative disorders and cancers.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty