Would you like to log in again?
Fibronectin Human ELISA, BioVendor
Supplier: BioVendor
RAF014R
102552-228EA 1054.98 USD InStock
102552-228
Fibronectin Human ELISA, BioVendor
Assays ELISAs
Fibronectin is an ubiquitous extracellular glycoprotein that exists in a soluble form in body fluids and in an insoluble form in the extracellular matrix.
  • Calibration Range: 0.31-20 ng/mL
  • Limit of Detection: 0.1 ng/mL
  • Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 50 uL/well
Fibronectin is an ubiquitous extracellular glycoprotein that exists in a soluble form in body fluids and in an insoluble form in the extracellular matrix. It plays a major role in many important physiological processes, such as embryogenesis, wound healing, hemostasis and thrombosis. Fibronectin is secreted as a dimer with a monomer molecular weight of 220–250 kDa. Fibronectin is composed of three types of modules organized into functional domains that are resistant to proteolysis and contain binding sites for extracellular matrix proteins, cell surface receptors and circulating blood proteins. In general, fibronectin is synthesized by and present around fibroblasts, endothelial cells, chondrocytes, glial cells, and myocytes. Fibronectin is abundant in the connective tissue matrix and in basement membranes. Extremely high levels of the glycoprotein are found in plasma. Fibronectin has been reported to play a major role in the cardiovascular system, being functionally involved in blood clotting, atherosclerosis, hypertension and myocardial infarction. Fibronectin´s importance during cancer progression has further been shown. It represents an active element in the process of T cell activation in the immune cascade triggered by organ transplantation. Fetal fibronectin has been described as a predictor of term labor.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty