Would you like to log in again?
GDF-15/MIC-1 Human ELISA, BioVendor
Supplier: BioVendor
RD191135200R
102552-234EA 694.78 USD InStock
102552-234
GDF-15/MIC-1 Human ELISA, BioVendor
Assays ELISAs
GDF-15/MIC-1 is synthesized as a 62-kDa precursor protein, which, after cleavage by furin-like protease, is secreted as 25-kDa disulfide-linked dimmer.
  • Calibration Range: 70-4480 ng/mL
  • Limit of Detection: 22 pg/mL
  • Plasma, Serum: 30 uL/well
GDF-15/MIC-1 is synthesized as a 62-kDa precursor protein, which, after cleavage by furin-like protease, is secreted as 25-kDa disulfide-linked dimmer. GDF-15/MIC-1 is weakly produced under baseline condition in most tissues such as brain, liver, kidney, pancreas, but not normally in many other organs including the heart, and is highly expressed in placenta and moderately in prostate. GDF-15 is upregulated by cardiovascular events triggering oxidative stress, including pressure overload, and atherosclerosis. Moreover, increased circulating GDF-15 concentrations have been linked to an enhanced risk of future adverse cardiovascular events in elderly woman and it was describe as a new biomarker of the risk of death in patients with non-ST-elevation acute coronary syndrome. Serum GDF-15 concentrations increase in maternal serum with advancing gestation in normal pregnancy. Low GDF-15 concentrations reportedly are associated with an increased risk of preterm labor or miscarriage.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty