Would you like to log in again?
Ghrelin Acylated Human Express ELISA, BioVendor
Supplier: BioVendor
RA194062400R
102552-236EA 1352.03 USD InStock
102552-236
Ghrelin Acylated Human Express ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of acylated ghrelin.
  • Calibration Range: 1.96 - 250 pg/ml
  • Limit of Detection: 0.3 pg/ml
  • Plasma-EDTA+PHMB: 20 µl/well
Ghrelin discovered in 1999, is fast becoming an endokrinology target of the millennium. Ghrelin, identifi ed in rat stomach as an endogenous ligand for the GH secretagogue receptor, is mainly produced in stomach, but has been demonstrated in many other organs [4, 5]. In addition to GHreleasing properties and its orexant action, Ghrelin could act as an hormone having effects on gastric motility (similarity with the peptide hormone motilin), acidic secretion, cardiovascular action, antiproliferative effects, pancreatic and glucose metabolism function, sleep [6, 7, 8]. This Enzyme Immunometric Assay (EIA) is based on a doubleantibody sandwich technique. The wells of the plate supplied are coated with a monoclonal antibody specific to the C-terminal part of Ghrelin. This antibody will bind to any Ghrelin introduced into the wells (standard or sample). The acetylcholinesterase (AChE) - Fab’ conjugate (Tracer) which recognises the N-terminal part of Acylated Ghrelin is also added to the wells. The two antibodies then form a sandwich by binding on different parts of the Acylated Ghrelin. The sandwich is immobilised on the plate so reagents in excess may be washed away. The concentration of Acylated Ghrelin (human) is determined by measuring the enzymatic activity of immobilized Tracer using Substrate Solution (Ellman’s Reagent). AChE Tracer acts on Substrate Solution (Ellman’s Reagent) to form a yellow compound that strongly absorbs at 414 nm.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty