Would you like to log in again?
Ghrelin Unacylated Mouse/Rat Express ELISA, BioVendor
Supplier: BioVendor
RA394063400R
102552-242EA 1352.03 USD InStock
102552-242
Ghrelin Unacylated Mouse/Rat Express ELISA, BioVendor
Assays ELISAs
Ghrelin stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis.
  • Calibration Range: 1.96-250 pg/mL
  • Limit of Detection: 0.6 pg/mL
  • Plasma-EDTA: 10uL/well
Ghrelin, an endogenous ligand for the growth hormone secretagogue receptor, is synthesized principally in the stomach
Ghrelin, an endogenous ligand for the growth hormone secretagogue receptor, is synthesized principally in the stomach. It stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis. The peptide consists of 28 amino acids, with a n-octanoylation of the serine-3 residue, which is necessary for the biological activity mentioned below. Ghrelin is present in the peripheral circulation under two forms: acylated and unacylated.
It stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis. The peptide consists of 28 amino acids, with a n-octanoylation of the serine-3 residue, which is necessary for the biological activity mentioned below. Ghrelin is present in the peripheral circulation under two forms: acylated and unacylated.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty