Would you like to log in again?
Glucagon Human ELISA, Multispecies specificity, BioVendor
Supplier: BioVendor
YK090
102552-246EA 1088.05 USD InStock
102552-246
Glucagon Human ELISA, Multispecies specificity, BioVendor
Assays ELISAs
This EIA kit has been developed by using polyclonal antibody against glucagon, synthetic glucagon as standard antigen and biotinylated glucagon as labeled antigen.
  • Calibration Range: 50-10,000 pg/mL
  • Plasma: 50-100 uL/well
According to many studies on glucagon immunoassay, it has been established that the antibody against the C – terminal fragment ( 19 – 29 ) of glucagon has specific binding with pancreatic glucagon, whereas the antibody against the N – terminal fragment ( 9 – 19 ) of glucagon has specific binding with both of pancreatic and intestinal glucagon ( total glucagon ). Once, 30K by Unger et. al had been widely used as an antibody specific for the C – terminal fragment of glucagon, but Nishino, Shima and Yanaihara et. al succeeded in producing pancreatic glucagon specific antibody using synthetic peptide with the C – terminal fragment ( 19 – 29 ) of glucagon as immunogen in 1981. This EIA kit has been developed by using polyclonal antibody against glucagon ( 19 – 29 ), synthetic glucagon as standard antigen and biotinylated glucagon as labeled antigen for the measurment of rat, mouse or human glucagon in plasma.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty