Would you like to log in again?
GM-CSF Human ELISA, BioVendor
Supplier: BioVendor
RAF016R
102552-250EA 1054.98 USD InStock
102552-250
GM-CSF Human ELISA, BioVendor
Assays ELISAs
GM-CSF is a small glycoprotein growth factor which stimulates the production and function of neutrophils, eosinophils and monocytes
  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 2.9 pg/mL
  • Amniotic fluid, Cell culture supernatant, Plasma, Serum: 50 uL/well
GM-CSF is a small glycoprotein growth factor which stimulates the production and function of neutrophils, eosinophils and monocytes. It can be produced by a wide variety of tissue types. The biological effects of GM-CSF are mediated through its binding to cell surface receptors, which appear to be widely expressed by thermatopoietic cells and also by some non-thermatopoietic cells, such as endothelial cells.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty