Would you like to log in again?
Growth Hormone Human ELISA, BioVendor
Supplier: BioVendor
RCD017R
102552-262EA 566.74 USD InStock
102552-262
Growth Hormone Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of GH.
  • Calibration Range: 1-50 ng/ml
  • Limit of Detection: 0.2 ng/ml
  • Serum: 25 ul/well
For the direct quantitative determination of growth hormone by enzyme immunoassayin human serum.
Human growth hormone (hGH) is a polypeptide of 191 amino acids secreted by the somatroph cells of the anterior pituitary. Growth hormone is principally a regulator of body growth and its metabolic effects are primarily anabolic. Some of its effects include promotion of protein conservation through its involvement in a wide range of protein synthesis mechanisms, enhancement of glucose transport and facilitation of glycogen storage. In addition, it induces the release of somatomedins (insulin-like growth factors), which further mediate the cascade of growth promoting actions.
The principle of the following enzyme immunoassay test follows a typical one-step capture or ‘sandwich’ type assay. The assay makes use of two highly specific monoclonal antibodies: A monoclonal antibody specific for hGH is immobilized onto the microwell plate and another monoclonal antibody specific for a different region of hGH is conjugated to horse radish peroxidase (HRP). hGH from the sample and standards are allowed to bind simultaneously to the plate and to the HRP conjugate. The washing and decanting steps remove any unbound HRP conjugate. After the washing step, the enzyme substrate is added. The enzymatic reaction is terminated by addition of the stopping solution. The absorbance is measured on a microtiter plate reader. The intensity of the color formed by the enzymatic reaction is directly proportional to the concentration of hGH in the sample. A set of standards is used to plot a standard curve from which the amount of hGH in subject samples and controls can be directly read.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty