Would you like to log in again?
sICAM-3 Human ELISA, BioVendor
Supplier: BioVendor
RAF105R
102552-274EA 1054.98 USD InStock
102552-274
sICAM-3 Human ELISA, BioVendor
Assays ELISAs
Intercellular Adhesion Molecule-3 (ICAM-3) is a member of the immunoglobulin supergene family and functions as a ligand for the Lymphocyte Function-Associated Antigen-1 (LFA-1).
  • Calibration Range: 0.78-50 ng/mL
  • Limit of Detection: 0.38 ng/mL
  • Amniotic fluid, Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 20 µL/well
Intercellular Adhesion Molecule-3 (ICAM-3) is a member of the immunoglobulin supergene family and functions as a ligand for the Lymphocyte Function-Associated Antigen-1 (LFA-1). Three counter-receptors have been described for LFA-1, intercellular adhesion molecule 1 (ICAM-1), ICAM-2 and ICAM-3. LFA-1, an alpha-beta complex, is a member of the leukocyte integrin family which mediate lymphocyte adhesion. ICAM-3 is a heavily glycosylated protein of 124 kDa with a polypeptide core of 57 kDa. The integral membrane protein with five immunoglobulin-like domains shares high homology to ICAM-1 and ICAM-2 in the extracellular region. In contrast to ICAM-1 and ICAM-2, ICAM-3 is absent on endothelia. ICAM-3 is expressed on resting lymphocytes, monocytes and neutrophils, representing the major LFA-1 ligand on these cells. The finding that adhesion of resting T lymphocytes to LFA-1 occurs primarily via ICAM-3 combined with the fact that ICAM-3 is much better expressed than other LFA-1 ligands on monocytes and resting lymphocytes implies an important role for ICAM-3 in the initiation of immune responses. ICAM-3 was found to be involved in the regulation of LFA-1/ ICAM-1 dependent leukocyte intercellular interactions. The initial interaction of ICAM-3 with LFA-1 might increase LFA-1-mediated cell binding to ICAM-1. Furthermore, ICAM-3 expression has been shown for dendritic epidermal Langerhans cells, whereas it is absent on other dendritic cells from different lymphoid organs. Thus potential function of ICAM-3 at the initiation phase of LC-leukocyte interactions taking place during skin localized immune reactions can be postulated. Recent data suggest that ICAM-3 expression can be induced on endothelial cells in lymphoid neoplasms as shown for Hodgkin's and non-Hodgkin's disease. ICAM-3 is a very interesting molecule involved in the initial immune response thus suggesting an important role as a disease marker for a number of different indications and pathological situations.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty