Would you like to log in again?
IFN-gamma Human ELISA, BioVendor
Supplier: BioVendor
RAF021R
102552-276EA 1054.98 USD InStock
102552-276
IFN-gamma Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of IFN-gamma.
  • Calibration Range: 1.6-100 pg/ml
  • Limit of Detection: 0.99 pg/ml
  • Cell culture supernatant, Plasma, Serum: 50 ul/well
The Human IFN-GAMMA ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of Human IFN-GAMMA. SUMMARY - IFN-GAMMA, also called type II interferon, is a homodimeric glycoprotein containing approximately 21 to 24 kD subunits. The human IFN-GAMMA gene, situated on chromosome 12, contains three introns, the four exons code for a polypeptide of 166 amino acids, 20 of which constitute the signal peptide.
An anti-human IFN-GAMMA coating antibody is adsorbed onto microwells. Human IFN-GAMMA present in the sample or standard binds to antibodies adsorbed to the microwells. A biotinconjugated anti-human IFN-GAMMA antibody is added and binds to Human IFN-GAMMA captured by the first antibody. Following incubation unbound biotin-conjugated antiHuman IFN-GAMMA antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotinconjugated anti-Human IFN-GAMMA antibody. Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty