Would you like to log in again?
IFN-omega Human ELISA, BioVendor
Supplier: BioVendor
RAF025R
102552-286EA 1054.98 USD InStock
102552-286
IFN-omega Human ELISA, BioVendor
Assays ELISAs
IFNω is a major component of human leukocyte IFN.
  • Calibration Range: 4.7-300 pg/mL
  • Limit of Detection: 1.5 pg/mL
  • Cell culture supernatant, Plasma, Serum: 25 uL/well
IFNω belongs to class I interferon. In the human genome, the IFNω gene family consists of seven members located on chromosome 9. IFNω is a major component of human leukocyte IFN. Therapeutically administered IFNω may cause measurable serum concentrations in the corresponding patients. Monitoring of these IFNω serum levels provides an important tool in therapy.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty