Would you like to log in again?
Immunoglobulin A, (Secretory), in Stool, IgA, Human ELISA, BioVendor
Supplier: BioVendor
IC6100
102552-288EA 926.26 USD InStock
102552-288
Immunoglobulin A, (Secretory), in Stool, IgA, Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of IgA.
  • Saliva, stool: 100 µl/well
The ImmuChrom ELISA Kit is intended for the quantitative determination of sIgA in stool and saliva. Secretory IgA is comprised of two immunglobuline A molecules, which are joined by a J-protein and a secretory component. The secretory component is synthesized by epithelial cells of the mucous membrane of gastrointestinal, respiratory and urogenitaltract. It is also produced by the saliva, tear and mammary glands. The plasma cells in the subendothelial area of mucous membranes are releasing a complex of two IgA-molecules, which are joined over the J-protein. This complex is binding to a secretory component, located at the surface of the epithel cell. After binding, the sIgA is transported across the cell and excreted by exocytosis. The determination of secretory IgA (sIgA) allows a first overview of the functionality of the gastrointestinal associated immune system (GALT). At this the secretory power and the degree of stimulation of the plasma cells of the intestinal submucosa is determined.
The sIgA-ELISA test determines human secretory IgA according to the 'sandwich'- principle. sIgA in sample, standard and controls binds to antibodies, which are coated to the microtiterplate. After a washing step a peroxidase labeled detection antibody is added. A second washing step is followed by the addition of the substrate which is converted to a colored product by the peroxidase. The reaction is terminated by the addition of an acidic stop solution. The optical densities are read at 450 nm (against the reference wavelength 620 nm) in a microtiterplate reader. The sIgA concentration can be calculated from the standard curve
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty