Would you like to log in again?
IGFBP-3 Total Human ELISA, BioVendor
Supplier: BioVendor
E03A
102552-294EA 896.88 USD InStock
102552-294
IGFBP-3 Total Human ELISA, BioVendor
Assays ELISAs
The development of specific immunoassays for IGFBP-3, recognizing the complete high molecular weight complex, provided new insights into ternary complex regulation.
  • Calibration Range: 0.4–30 ng/mL
  • Limit of Detection: 0.1 ng/mL
  • Cerebrospinal fluid, Plasma-Heparin, Serum
Insulin-like growth factors (IGF)-I and -II are bound to specific binding proteins (IGFBPs) in the circulation. To date, at least six binding proteins can be distinguished on the basis of their amino acid sequence. They are designated as IGFBP-1, IGFBP-2 to IGPBP-6. Lately the discovery of a new IGFBP-7 has been discussed. The predominating IGFBP in blood is IGFBP-3, which largely determines the total IGF-I and IGF-II concentration. In contrast to the other binding proteins, IGFBP-3 has the unique property to associate with an acid-labile non-binding subunit (ALS) after binding of either IGF-I or IGF-II. Most of the IGFBP-3 in plasma is present as the high molecular weight ternary complex, however, small amounts of free IGFBP-3 are also found. The development of specific immunoassays for IGFBP-3, recognizing the complete high molecular weight complex, provided new insights into ternary complex regulation. Several factors besides GH influence IGFBP-3 levels: age including sexual development, nutrition, hypothyroidism, diabetes mellitus, liver function and kidney function. Measurement over 24 hours revealed constant circadian levels.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty