Would you like to log in again?
IGF-2 Human ELISA, BioVendor
Supplier: BioVendor
E30
102552-296EA 915.29 USD InStock
102552-296
IGF-2 Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of IGF-2.
  • Calibration Range: 0.02 to 3600 ng/ml
  • Limit of Detection: 0.02 ng/ml
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum
The insulin-like growth factors (IGF)-1 and –2 play a pivotal role in the regulation of proliferation and differentiation of several tissue types. IGF-1 also called Somatomedin C has a molecular weight of 7.469 kDa. Its expression is mainly regulated by Growth Hormone and nutrition. But several hormones and peptide factors are known to influence IGF-2 synthesis in different tissues. Bioavailability of the IGFs is regulated by specific binding proteins (IGFBP). Beside the high affinity Insulin-like Growth Factor Binding Proteins 1–6, IGFs are also bound be IGFBP-related Proteins. These binding proteins bind IGF-1 and IGF-2 with the same affinity or prefer IGF-2. Direct measurement of IGFs in serum samples without pretreatment results in false values because of the extremely slow dissociation of the IGF/IGFBP complexes during the assay incubation only a part of the IGF-2 in the specimen can bind to the antibodies and be detected. Therefore, various techniques were applied to physically separate IGF-2 from its binding proteins before measurement, including (a) size exclusion chromatography under acidic conditions, (b) solid-phase extraction and © acid-ethanol extraction. These techniques, however, are either inconvenient or time-consuming or give incomplete and not-reproducible recoveries. This assay is easy, fast and results do not depend on the binding protein concentration of the sample. Its based on the high specificity of the employed antibodies for IGF-2. There is virtually no cross-reactivity with IGF-1. This allows the separation of IGF-2 from the binding proteins by acidification and blocking of the free binding proteins with IGF-1. Thus, the endogenous IGF-2 is free in solution.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty