Would you like to log in again?
Interleukin-12 (p70) Human ELISA, BioVendor
Supplier: BioVendor
RAF030R
102552-308EA 1054.98 USD InStock
102552-308
Interleukin-12 (p70) Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of Interleukin-12 (p70).
  • Calibration Range: 15.6-1000 pg/ml
  • Limit of Detection: 5 pg/ml
  • Cell culture supernatant, Plasma, Serum: 50 ul/well
The Human IL-12 p70 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human IL-12p70. SUMMARY - Interleukin-12 (IL-12) is a pleiotropic cytokine, formerly termed cytotoxic lymphocyte maturation factor (CLMF) or natural killer cell stimulatory factor (NKSF), which is produced primarily by stimulated macrophages. It was originally identified as a factor produced by human EpsteinBarr Virus transformed B cell lines. Meanwhile IL-12 has been shown to be a proinflammatory cytokine produced by phagocytic cells, B cells, and other antigen-presenting cells that modulate adaptive immune responses by favoring the generation of T-helper type 1 cells. An anti-human IL-12p70 coating antibody is adsorbed onto microwells.
Human IL-12p70 present in the sample or standard binds to antibodies adsorbed to the microwells. A biotin-conjugated anti-human IL-12p70 antibody is added and binds to human IL-12 p70 captured by the first antibody. Following incubation unbound biotin-conjugated anti-human IL-12p70 antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotinconjugated anti-human IL-12p70 antibody. Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of human IL-12p70 present in the sample or standard. The reaction is terminated by addition of acid and absorbance is measured at 450 nm. A standard curve is prepared from 7 human IL-12p70 standard dilutions and human IL-12p70 sample concentration determined.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty