Would you like to log in again?
Interleukin-12, p70, IL-12, Mouse ELISA, BioVendor
Supplier: BioVendor
RAF031R
102552-310EA 1014.71 USD InStock
102552-310
Interleukin-12, p70, IL-12, Mouse ELISA, BioVendor
Assays ELISAs
IL-12 is a heterodimeric cytokine which shares the p40 subunit.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 10 pg/mL
  • Body fluids, Cell culture supernatant, Plasma, Serum: 50 uL/well
Cytokines and chemokines and related molecules, Oncology
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty