Would you like to log in again?
Interleukin-12/Interleukin-23 p40 Human ELISA, BioVendor
Supplier: BioVendor
RAF034R
102552-316EA 1055.05 USD InStock
102552-316
Interleukin-12/Interleukin-23 p40 Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of Interleukin-12/Interleukin-23 p40.
  • Calibration Range: 31.3-2000 pg/ml
  • Limit of Detection: < 10 pg/ml
  • Cell culture supernatant, Plasma, Serum: 50 ul/well
The human IL-12/IL-23 p40 ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human IL-12/IL-23 p40. The human IL-12/IL-23 p40 ELISA is for research use only. Not for diagnostic or therapeutic procedures.
IL-23 and IL-12 are heterodimeric cytokines which share the p40 subunit, but have unique second subunits, IL-23p19 and IL-12p35.
An anti-human IL-12/IL-23 p40 coating antibody is adsorbed onto microwells. Human IL-12/IL-23 p40 present in the sample or standard binds to antibodies adsorbed to the microwells. A biotin-conjugated anti-human IL-12/IL23 p40 antibody is added and binds to human IL12/IL-23 p40 captured by the first antibody. Following incubation unbound biotin-conjugated antihuman IL-12/IL-23 p40 antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotin-conjugated anti-human IL-12/IL-23 p40 antibody. Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of human IL-12/IL-23 p40 present in the sample or standard. The reaction is terminated by addition of acid and absorbance is measured at 450 nm. A standard curve is prepared from 7 human IL-12/IL-23 p40 standard dilutions and human IL12/IL-23 p40 sample concentration determined.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty