Would you like to log in again?
Interleukin-17A, IL-17A, Human ELISA, BioVendor
Supplier: BioVendor
RAF039R
102552-328EA 1055.05 USD InStock
102552-328
Interleukin-17A, IL-17A, Human ELISA, BioVendor
Assays ELISAs
Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism.
  • Calibration Range: 1.6-100 pg/mL
  • Limit of Detection: 0.5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-Heparin, Serum: 50 uL/well
Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism. IL-17 expression in airways is upregulated upon allergen exposure such as in allergic asthma. Its crucial role in T-cell activation suggests that IL-17 is involved in the development of autoimmune arthritis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty