Would you like to log in again?
Interleukin-17AF, IL-17AF, Human ELISA, BioVendor
Supplier: BioVendor
RAF041R
102552-330EA 1055.05 USD InStock
102552-330
Interleukin-17AF, IL-17AF, Human ELISA, BioVendor
Assays ELISAs
Of the six IL-17 family members, IL-17F and IL-17A share the strongest homology.
  • Calibration Range: 31.3-2000 pg/mL
  • Limit of Detection: 8.8 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-Heparin, Serum: 50 uL/well
IL-17A and IL-17F are members of the IL-17 family and signature Th17 cytokines. Of the six IL-17 family members, IL-17F and IL-17A share the strongest homology (50% amino acid identity) and the two genes are located in the same chromosomal region. Although both IL-17A and IL- 17F were originally found to be produced as disulfide-linked homodimers, recent studies have confirmed the production of IL-17A/IL-17F heterodimers in in vitro-differentiated and polarized Th17 cells. Activated human CD4+ T cells in culture were found to secrete IL-17F homodimer at 10-fold higher levels than IL-17A homodimer, suggesting that the majority of the IL-17A protein expressed exists in the form of the IL-17AF heterodimer. Studies of Th17 polarized mouse splenocytes also indicate the majority of IL-17 made is the IL-17A/F heterodimer. IL-17F and IL-17A have been observed in tissue samples from various autoimmune diseases, such as rheumatoid arthritis, multiple sclerosis, psoriasis, inflammatory bowel disease, and asthma.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty