Would you like to log in again?
Interleukin-1 alpha, IL-1a, Rat ELISA, BioVendor
Supplier: BioVendor
RAF047R
102552-346EA 1015.68 USD InStock
102552-346
Interleukin-1 alpha, IL-1a, Rat ELISA, BioVendor
Assays ELISAs
IL-1α is predominantly a cell-associated, non-secreted proinflammatory cytokine.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 4 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well
IL-1α is predominantly a cell-associated, non-secreted proinflammatory cytokine. It is produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. High levels of IL-1α are associated with sepsis, rheumatoid arthritis, inflammatory bowel disease, acute and chronic myelogenous leukemia, insulin-dependent diabetes mellitus, and atherosclerosis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty