Would you like to log in again?
Interleukin-1 beta, IL-1b, Rat ELISA, BioVendor
Supplier: BioVendor
RAF050R
102552-352EA 1015.72 USD InStock
102552-352
Interleukin-1 beta, IL-1b, Rat ELISA, BioVendor
Assays ELISAs
IL-1b has been shown to induce fever and is thought to contribute to wasting of muscles.
  • Calibration Range: 31.3-2000 pg/mL
  • Limit of Detection: 4 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well
Interleukin-1 (IL-1), originally described in 1972 as lymphocyte activating factor (LAF) for its effects on thymocytes, is a polypeptide cytokine with two molecular forms. Both forms appear to mediate identical ranges of biological activity which include synthesis of the acute phase proteins by hepatocytes, chemotaxis of polymorphonucleocytes, and release of polymorphonucleocytes from blood and bone marrow. These effects coined the acronym leukocyte endogenous mediator (LEM). Early researchers also called IL-1b endogenous pyrogen, and it has been shown to induce fever and is thought to contribute to wasting of muscles (PIF, proteolysis inducing factor). Other activities associated with IL-1 are the induction of Prostaglandin E2 by synovial cells and release of collagenase with resulting destruction of cartilage and bone resorption (catabolin, osteoclast activation factor). In addition, IL-1, has multiple immunological functions including enhancement of IL-2 production by T cells and activation of B-cells (BAF) and thymocytes. A true pleiotrope, IL-1 may have tumoricidal activity via its release of IL-2 and interferon gamma and be indirectly antiviral by stimulating fibroblasts to release interferon beta. In its role as mediator of sepsis, IL-1 has most recently been described as enhancing the growth of virulent E.coli. The two distinct molecular forms of IL-1 are thought to be derived from two genes. After transcription, as 31 kD precursor polypeptide is cleaved to give rise to mostly cell membrane associated IL-1a and secreted IL-1b. Both have the same molecular weight of 15 kD but have different isoelectric points of 5 and 7, respectively. Despite sequence homology of only 20%, both forms are thought to bind to the same receptor. IL-1 inhibitors that vary only in their degree of glycosylation have been described to bind to the IL-1 receptor. These inhibitors are structurally related to IL-1b and may be important in regulation of IL-1b action. Low levels of IL-1b have been reported in normal serum. It is thought that IL-1 genes are induced to respond to tissue damage or in infection. Elevated levels have been reported in a number of infectious disease conditions and in noninfectious inflammatory conditions such as Crohn’s disease. In addition to elevated serum levels, IL-1 has been found in synovial fluids of patients with rheumatoid arthritis and in cerebrospinal fluid after neurological inflammation or insult. At the other end of the spectrum, low levels of IL-1 have been found in malnutrition and advanced neoplasia suggesting perhaps a complex immunological and physiological regulatory role for this cytokine.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty