Would you like to log in again?
Interleukin-2, IL-2, Mouse ELISA, BioVendor
Supplier: BioVendor
RAF053R
102552-358EA 1014.71 USD InStock
102552-358
Interleukin-2, IL-2, Mouse ELISA, BioVendor
Assays ELISAs
Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens.
  • Calibration Range: 109-7000 pg/mL
  • Limit of Detection: 30 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 50 uL/well
Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens. IL-2 plays a pivotal role in for the expansion of most T-cells, natural killer cells and B-cells during certain phases of their response. IL-2 gene expression is regulated at the transcriptional level by several activation pathways. Antigen-specific proliferation of helper and cytotoxic T-lymphocytes following stimulation is critically dependent on IL-2 expression, secretion, and binding to receptors for IL-2 induced in an autocrine fashion on the surface of T-cells. Apart from its most important role to mediate antigen-specific T-lymphocyte proliferation, IL-2 modulates the expression of IFN-g and major histocompatibility antigens, stimulates proliferation and differentiation of activated B-cells, augments natural killer cell activity and inhibits granulocyte-macrophage colony formation. Alterations in the ability of T-cells to synthesize IL-2 have been observed in physiologic and pathologic states.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty