Would you like to log in again?
Interleukin-21, IL-21, Human ELISA, BioVendor
Supplier: BioVendor
RAF057R
102552-364EA 1054.98 USD InStock
102552-364
Interleukin-21, IL-21, Human ELISA, BioVendor
Assays ELISAs
Interleukin-21 is the most recently discovered member of the type-I cytokine family.
  • Calibration Range: 78-5000 pg/mL
  • Limit of Detection: 20 pg/mL
  • Cell culture supernatant, Plasma, Plasma-Citrate, Plasma-EDTA: 50 uL/well
Interleukin-21 is the most recently discovered member of the type-I cytokine family. Structurally, IL-21 shows homology to IL-2, IL-4, and IL-15 proteins. IL-21 shares the common gamma-chain with these three cytokines, but, in addition, binds to a unique IL-21R alpha chain, and activates the JAK/STAT pathway. IL-21 has pleiotropic actions, from augmenting the proliferation of T cells and driving the differentiation of B cells into memory cells and terminally differentiated plasma cells to augmenting the activity of natural killer cells. IL-21 is mainly produced by activated T-cells, but targets a broad range of lymphoid and myeloid cells of the immune system and therefore is able to regulate innate and acquired immune responses. IL-21 has pathologic function in immune-inflammatory diseases. Data suggest the existence of a positive autocrine loop that could help to amplify and stabilize IL-21-driven, T cell-mediated responses. Moreover, it has antitumour activity and may have a role in the development of autoimmunity. IL-21 may have therapeutic potentials as an antitumor agent in the clinic.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty