Would you like to log in again?
sInterleukin-2 Receptor, sIL-2R, Human ELISA, BioVendor
Supplier: BioVendor
RAF106R
102552-374EA 1054.98 USD InStock
102552-374
sInterleukin-2 Receptor, sIL-2R, Human ELISA, BioVendor
Assays ELISAs
sIL-2R is a marker for T cell activation.
  • Calibration Range: 0.31-20 ng/mL
  • Limit of Detection: 0.27 ng/mL
  • Cell culture supernatant, Plasma, Serum: 50 µL/well
The receptor of the cytokine IL-2 plays a crucial role in the regulation of the immune response. Binding of IL-2 to its receptor (IL-2R) on the surface of T-lymphocytes triggers a series of intracellular signaling events that result in the activation and proliferation of resting T cells and in the generation of helper, suppressor and cytotoxic T cells, which mediate immune reactions. sIL-2R is a marker for T cell activation. It can be an indicator of a wide spectrum of disorders involving immune activation. IL2R can be found at significantly elevated levels in a broad range of disorders such as tumor diseases, autoimmune diseases (e.g. sarcoidosis), organ rejection and various infections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty