Would you like to log in again?
Interleukin-6, IL-6, Rat ELISA, BioVendor
Supplier: BioVendor
RAF072R
102552-392EA 1015.54 USD InStock
102552-392
Interleukin-6, IL-6, Rat ELISA, BioVendor
Assays ELISAs
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms.
  • Calibration Range: 31-2000 pg/mL
  • Limit of Detection: 12 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms. IL-6 is usually not produced constitutively by normal cells, but its expression is readily induced by a variety of cytokines, lipopolysaccharide or viral infections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty