Would you like to log in again?
sInterleukin-6 Receptor, sIL-6R, Human ELISA, BioVendor
Supplier: BioVendor
RAF107R
102552-396EA 1055.05 USD InStock
102552-396
sInterleukin-6 Receptor, sIL-6R, Human ELISA, BioVendor
Assays ELISAs
Interleukin-6 (IL-6) is a multifunctional cytokine involved in the regulation of the immune response, hematopoiesis and acute phase response.
  • Calibration Range: 0.08-5 ng/mL
  • Limit of Detection: 0.01 ng/mL
  • Amniotic fluid, Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 20 µL (1:50 prediluted)
Interleukin-6 (IL-6) is a multifunctional cytokine involved in the regulation of the immune response, hematopoiesis and acute phase response. It has been recognized to be a member of the a-helical cytokine family. IL-6 exerts its action via a cell surface receptor which consists of two subunits, an 80 kDa ligand binding subunit (gp80) of 468 amino acids and a 130 kDa signal transducing protein (gp130) of 896 amino acid residues. The cDNAs of both proteins have been cloned. Both subunits belong to the recently recognized hematopoietic receptor superfamily which includes many cytokine receptors. Characterisation of the extracellular portion of the 80 kDa IL-6 receptor revealed the existence of a single immunoglobulin-like domain in the NH2-terminal of the extracellular region, which does not contribute to ligand binding. The remainder of the extracellular domain however is essential for low affinity ligand binding, which consecutively triggers the association of the receptor and gp130 thus forming a high affinity binding site for IL-6. For many cytokine receptors soluble forms have been demonstrated. These soluble molecules have been observed to retain ligand binding capacity and therefore compete with the membrane receptors, thus acting as antagonists. A soluble form of the Human gp80 protein has been detected in serum and urine samples. This 55 kDa protein representing the extracellular portion of gp80 is generated by shedding, a process that seems to be controlled by protein kinase C. It is still functional, indicating that soluble gp80 plays a biological role in promoting IL-6 activity. So far, the soluble IL-6 receptor is unique in acting as an agonist together with its ligand. The role of soluble IL-6R as a marker for a number of pathological situations has been demonstrated.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty